5-Brom-2-thiophencarbonitril
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
5-Bromo-2-thiophenecarbonitrile, 5-Bromothiophene-2-carbonitrile, 2-Cyano-5-bromothiophene, 2-Bromo-5-cyanothiophene
AS33746G1
AS33746G5
ACTIAS33746G1EA
0
CHF
ACTIAS33746G1
ACTIAS33746G5
5-Brom-2-thiophencarbonitril
5-Brom-2-thiophencarbonitril
Formel:
C₅H₂BrNS MW: 188,0500 g/mol |
MDL Number:
MFCD04974080 CAS-Nummer: 2160-62-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...