2-Chlor-5-methylnicotinsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Chloro-5-methylpyridine-3-carboxylic acid

AS32615G1 AS32615G5
ACTIAS32615G1EA 0 CHF
ACTIAS32615G1 ACTIAS32615G5
2-Chlor-5-methylnicotinsäure
2-Chlor-5-methylnicotinsäure
Formel: C₇H₆ClNO₂
MDL Number: MFCD07375371
CAS-Nummer: 66909-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR