2-Chlor-6-hydroxynicotinsäure

Lieferant: ACTIVATE SCIENTIFIC

AS31267G1 AS31267G5
ACTIAS31267G1EA 0 CHF
ACTIAS31267G1 ACTIAS31267G5
2-Chlor-6-hydroxynicotinsäure
2-Chlor-6-hydroxynicotinsäure
Formel: C₆H₄ClNO₃
MW: 173,5500 g/mol
MDL Number: MFCD09757508
CAS-Nummer: 38025-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR