3,5-Dibrompyridin-N-oxid
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3,5-Dibromopyridine1-oxide
AS34902G1
AS34902G5
ACTIAS34902G1EA
0
CHF
ACTIAS34902G1
ACTIAS34902G5
3,5-Dibrompyridin-N-oxid
3,5-Dibrompyridin-N-oxid
Formel:
C₅H₃Br₂NO MW: 252,8914 g/mol |
MDL Number:
MFCD00160750 CAS-Nummer: 2402-99-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...