3,5-Dimethyl-4-methoxyphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Methoxy-3,5-dimethylphenylboronic acid, 4-Methoxy-3,5-dimethylbenzeneboronic acid

AS50361G100 AS50361G5
ACTIAS50361G100EA 0 CHF
ACTIAS50361G100 ACTIAS50361G5
3,5-Dimethyl-4-methoxyphenylboronsäure
3,5-Dimethyl-4-methoxyphenylboronsäure
Formel: C₉H₁₃BO₃
MDL Number: MFCD01114648
CAS-Nummer: 301699-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR