2-Chlor-4-methylphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS32681G1EA 0 CHF
ACTIAS32681G1 ACTIAS32681G5
2-Chlor-4-methylphenylboronsäure
2-Chlor-4-methylphenylboronsäure
Formel: C₇H₈BClO₂
MW: 170,40 g/mol
MDL Number: MFCD03411936
CAS-Nummer: 145349-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR