4-(Hydroxymethyl)phenylboronsäurepinakolester
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl alcohol, 2-[4-(Hydroxymethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-(Hydroxymethyl)benzeneboronic acid pinacol ester, (4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol, 4-(Hydroxymethyl)phenylboronic acid Pinacol ester
AS50664G1
AS50664G5
ACTIAS50664G1EA
0
CHF
ACTIAS50664G1
ACTIAS50664G5
4-(Hydroxymethyl)phenylboronsäurepinakolester
4-(Hydroxymethyl)phenylboronic acid pinacol ester
Formel:
C₁₃H₁₉BO₃ MW: 234,10 g/mol |
MDL Number:
MFCD09837617 CAS-Nummer: 302348-51-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...