2-Chlor-4-fluorphenylhydrazin Hydrochlorid
Lieferant: ACTIVATE SCIENTIFIC
Synonyms: (2-Chloro-4-fluorophenyl)hydrazine hydrochloride
AS32388G1 AS32388G10 AS32388G5
ACTIAS32388G1EA 0 CHF
ACTIAS32388G1 ACTIAS32388G10 ACTIAS32388G5
2-Chlor-4-fluorphenylhydrazin Hydrochlorid
2-Chlor-4-fluorphenylhydrazin Hydrochlorid
Formel: C₆H₇Cl₂FN₂
MDL Number: MFCD03788524
CAS-Nummer: 497959-29-2