2-Formyl-5-methoxyphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Methoxy-2-formylphenylboronic acid

AS50866G1 AS50866G5
ACTIAS50866G1EA 0 CHF
ACTIAS50866G1 ACTIAS50866G5
2-Formyl-5-methoxyphenylboronsäure
2-Formyl-5-methoxyphenylboronsäure
Formel: C₈H₉BO₄
MW: 179,9657 g/mol
MDL Number: MFCD03001335
CAS-Nummer: 40138-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR