2-Formyl-4-methoxyphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

AS50865G1 AS50865G5
ACTIAS50865G1EA 0 CHF
ACTIAS50865G1 ACTIAS50865G5
2-Formyl-4-methoxyphenylboronsäure
2-Formyl-4-methoxyphenylboronsäure
Formel: C₈H₉BO₄
MW: 179,9657 g/mol
MDL Number: MFCD01319043
CAS-Nummer: 139962-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR