3-Amino-3-(4-bromphenyl)propionsäure

Lieferant: ACTIVATE SCIENTIFIC

AS24042G1 AS24042G25 AS24042G5
ACTIAS24042G1EA 0 CHF
ACTIAS24042G1 ACTIAS24042G25 ACTIAS24042G5
3-Amino-3-(4-bromphenyl)propionsäure
3-Amino-3-(4-bromphenyl)propionsäure
Formel: C₉H₁₀BrNO₂
MW: 244.0900 g/mol
MDL Number: MFCD00187112
CAS-Nummer: 39773-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR