3'-Chlor-4'-hydroxyacetanilid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Chloro-4-hydroxyacetanilide, N-(3-Chloro-4-hydroxyphenyl)acetamide

AS33980G1 AS33980G5
ACTIAS33980G1EA 0 CHF
ACTIAS33980G1 ACTIAS33980G5
3'-Chlor-4'-hydroxyacetanilid
3'-Chlor-4'-hydroxyacetanilid
Formel: C₈H₈ClNO₂
MW: 185,6100 g/mol
MDL Number: MFCD01318123
CAS-Nummer: 3964-54-3
EINECS: 223-570-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR