Ethyl-2,4-dimethylthiazol-5-carboxylat
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
Ethyl 2,4-dimethylthiazole-5-carboxylate
AS35139G1
AS35139G5
ACTIAS35139G1EA
0
CHF
ACTIAS35139G1
ACTIAS35139G5
Ethyl-2,4-dimethylthiazol-5-carboxylat
Ethyl-2,4-dimethylthiazol-5-carboxylat
Formel:
C₈H₁₁NO₂S MW: 185.24 g/mol |
MDL Number:
MFCD00052874 CAS-Nummer: 7210-77-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...