2,4-Dichlor-5-cyanthiazol

Lieferant: ACTIVATE SCIENTIFIC

AS33372G1 AS33372G5
ACTIAS33372G1EA 0 CHF
ACTIAS33372G1 ACTIAS33372G5
2,4-Dichlor-5-cyanthiazol
2,4-Dichlor-5-cyanthiazol
Formel: C₄Cl₂N₂S
MW: 179,0300 g/mol
MDL Number: MFCD08669569
CAS-Nummer: 82554-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR