5-Iod-2,4-dimethoxypyrimidin

Lieferant: ACTIVATE SCIENTIFIC

AS32400G1 AS32400G10 AS32400G5
ACTIAS32400G1EA 0 CHF
ACTIAS32400G1 ACTIAS32400G10 ACTIAS32400G5
5-Iod-2,4-dimethoxypyrimidin
5-Iod-2,4-dimethoxypyrimidin
Formel: C₆H₇IN₂O₂
MW: 266.04 g/mol
MDL Number: MFCD00090865
CAS-Nummer: 52522-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR