2-Chlorpyrimidin-4-carbonitril

Lieferant: ACTIVATE SCIENTIFIC

AS1151G1 AS1151G5
ACTIAS1151G1EA 0 CHF
ACTIAS1151G1 ACTIAS1151G5
2-Chlorpyrimidin-4-carbonitril
2-Chlorpyrimidin-4-carbonitril
Formel: C₅H₂ClN₃
MW: 139,5400 g/mol
MDL Number: MFCD00234989
CAS-Nummer: 75833-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR