1-Phenyl-1H-pyrazol-4-carbaldehyd

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-Phenylpyrazole-4-carboxaldehyde, 1-Phenyl-1H-pyrazole-4-carboxaldehyde

AS1615G1 AS1615G5
ACTIAS1615G1EA 0 CHF
ACTIAS1615G1 ACTIAS1615G5
1-Phenyl-1H-pyrazol-4-carbaldehyd
1-Phenyl-1H-pyrazol-4-carbaldehyd
Formel: C₁₀H₈N₂O
MW: 172,18 g/mol
MDL Number: MFCD02179568
CAS-Nummer: 54605-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR