Methyl-2-pyrazincarboxylat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 2-pyrazinecarboxylate, Methyl pyrazine-2-carboxylate

AS50271G100 AS50271G5
ACTIAS50271G100EA 0 CHF
ACTIAS50271G100 ACTIAS50271G5
Methyl-2-pyrazincarboxylat
Methyl-2-pyrazincarboxylat
Formel: C₆H₆N₂O₂
MW: 138.13 g/mol
MDL Number: MFCD00014611
CAS-Nummer: 6164-79-0
EINECS: 228-198-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR