Methyl-5-brompyrazin-2-carboxylat

Lieferant: ACTIVATE SCIENTIFIC

AS30971G1 AS30971G5
ACTIAS30971G1EA 0 CHF
ACTIAS30971G1 ACTIAS30971G5
Methyl-5-brompyrazin-2-carboxylat
Methyl-5-brompyrazin-2-carboxylat
Formel: C₆H₅BrN₂O₂
MDL Number: MFCD09991715
CAS-Nummer: 210037-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR