(5-Chlor-3-pyridinyl)methanol

Lieferant: ACTIVATE SCIENTIFIC

AS30513G1 AS30513G5
ACTIAS30513G1EA 0 CHF
ACTIAS30513G1 ACTIAS30513G5
(5-Chlor-3-pyridinyl)methanol
(5-Chlor-3-pyridinyl)methanol
Formel: C₆H₆ClNO
MDL Number: MFCD09743969
CAS-Nummer: 22620-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR