N,N-Dimethylformamid-[D7] (99.5% D)
Lieferant: CAMBRIDGE ISOTOPE LABS., INC.
|
Gefahr
|
Synonyms:
N,N-Dimethylmethansäureamid-D7
, DMF-D7
, N,N-Di[2H3]methyl[2H]formamid
, Ameisensäuredimethylamid-D7
CAMBDLM-25-10X1EA
0
CHF
CAMBDLM-25-10X1
N,N-Dimethylformamid-[D7] (99.5% D)
N,N-Dimethylformamid-[D7]
Formel:
DCOON(CD₃)₂ MW: 80,04 g/mol Siedepunkt: 152 °C (1013 hPa) Melting Pt: –60 °C Dichte: 1,03 g/cm³ (20 °C) Flash-Pt: 58 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00003286 CAS-Nummer: 4472-41-7 EINECS: 224-745-8 UN: 2265 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...