1,4,5,8-Tetraamino-9,10-anthrachinon 30% (Farbstoffgehalt), Sigma-Aldrich®
Lieferant: SIGMA-ALDRICH MICROSCOPY
Gefahr
215643-5GEA 121 CHF
215643-5G
1,4,5,8-Tetraamino-9,10-anthrachinon 30% (Farbstoffgehalt), Sigma-Aldrich®
1,4,5,8-Tetraamino-9,10-anthrachinon
Disperse blue 1 is a grey to black powder. Disperse dyes are known to be insoluble in water and are applied to the substrate as suspensions or dispersions by dissolving in hydrophobic regions of the polymers making up the fiber.
Formel: C₁₄H₁₂N₄O₂
MW: 268,27 g/mol
Melting Pt: 332 °C
Lager Temperatur: Room Temperature
MDL Number: MFCD00001225
CAS-Nummer: 2475-45-8
EINECS: 219-603-7
Order Now
Spezifikation Testergebnisse
Appearance Grey to black powder
Infrared spectrum Conforms to structure
UV-visible spectrum 0.01 g/L in 50% ethanol w/1 drop 1N NaOH