Mounting Bracket Kit for In-Line Separators, 1/2" to 3/4" Inlet/Outlet
Lieferant: Avantor Fluid Handling
VWR Item#MFLX29995-50
Your Organization's Price:
Optimize performance of your Liquid-Solids Separator.
  • Keep your Separators running properly with these kits
Choose from a wide range of replacement parts and accessories to ensure top performance of your Liquid-Solids Separator / Concentrator Units. Purge kits are available in manual or automatic forms so solids may be purged or bled as needed.
Mounting bracket kits are designed to support the separator at a near-upright position.