Guzzler 400-H Diaphragm hand pump, 8.5 GPM, 7 Strokes/gallon
Lieferant: Avantor Fluid Handling
VWR Item#MFLX07090-10
Your Organization's Price:
Low-cost, high-flow pumping solution.
  • Ideal for pumping shear-sensitive or particulate-laden fluids
  • Durable, portable pumps mount in any position
  • Available in cast aluminum or nonclogging Delrin® acetal plastic
  • 07090-09 has inlet and outlet on same side
Guzzler® hand pumps are designed for quick, easy fluid transfer.<br /><br />Order service kits, noncollapsing wire-reinforced vinyl hoses, stainless steel hose clamps, and check valves separately in table.