C-Flex Transfer Tubing, C-Flex® TPE Braided, 5/8" ID x 1" OD; 15 Ft
Lieferant: Avantor Fluid Handling
VWR Item#MFLX06433-09
Your Organization's Price:
Get more strength than standard C-Flex.
  • Ideal for general-purpose, single-use, and extended fluid path applications
<B>Applications:</B> For use with cell cultures, vessel and tank transfer, pump applications and cold storage. <br /><B>Characteristics: </B>C-Flex® formula 082, clear, animal derived component free (ADCF) resin and reinforced with polyester braid. <br /><B>Certifications: </B>Complies with USP Class VI standards.<br /><B>Temperature range:</B> -50 to 275 °F (-45 to 135 °C) <br /><B>Sterilize:</B> Autoclavable and gamma sterilizable.