oasig™ Lyophilised master mixes
Lieferant: PRIMERDESIGN
ZOASIGONEST150
ZOASIGONEST150EA 0 CHF
ZOASIGONEST150
oasig™ Lyophilised master mixes
Nukleinsäure-Reagenzien qPCR/RT-qPCR Enzyme und Kits
The oasig freeze drying process stabilises all of the active components allowing these reagents to be shipped and stored at room temperature. This hugely simplifies the logistics of purchasing, shipping and using the technology. Whether you are in a sophisticated laboratory in Texas or a mobile field hospital in Timbuktu we can supply complete qPCR kit and reagent packages to your door quickly and cheaply via standard shipping methods without the need for dry ice or a cold chain of any sort.
Freeze dried qPCR reagents stored and shipped at ambient temperatures:
  • No cold chain shipping
  • Simple transport logistics
  • Exceptional data quality
  • Simple protocol
  • Suitable for all real-time PCR machines
The performance of the reagents is second to none. We are confident that you will find excellent data quality and even see an improvement in data quality versus many traditional frozen master mixes.
Zertifizierungen: Primerdesign real-time PCR reagents are manufactured to the highest standards within our ISO9001:2008 and ISO13485:2012 certified quality management laboratory environment.
Verpackung: Kit contains 3 x 50 reaction vials and resuspension buffer