Viral inactivation solution, ultrareines Bioreagenz, J.T.Baker®

Lieferant: Avantor

X191-04EA 265 CHF
X191-04 X191-01 X191-09
Viral inactivation solution, ultrareines Bioreagenz, J.T.Baker®
Viral inactivation solution

J.T.Baker® Viral Inactivation Solution Ultrapure bioreagent is a highly effective, ready-to-use product, specifically developed for detergent-based viral inactivation in biopharmaceutical operations and plasma derived therapy processes. J.T.Baker® Viral Inactivation Solution is formulated with non-ionic detergent, that is readily biodegradable and alkylphenol ethoxylate free, addressing environmental concerns associated with REACH. This innovative solution is optimized to have low protein reactive impurities, low viscosity and compatibility with typical downstream processes. J.T.Baker® Viral Inactivation Solution Ultrapure bioreagent is available in a variety of sizes to support development to large scale commercial batches. Detail description provided in technical literature.


  • High purity, tested for bioburden and impurity
  • Effective inactivation of virus
  • Easily cleared in downstream processing, detectable to 1ppm
  • Readily biodegradable
  • 0.2 µm filtered solution


Proprietary Formulation. Not hazardous according to regulation 29 CFR 1910.1200 (OSHA HCS)

Order Now

Spezifikation Testergebnisse

Identification (by FTIR) Passes Test
Appearance Passes Test
Purity by HPLC min 95%
Assay Passes Test
Acid value max 10
Viscosity (25°C) For information only
Density 0.9 – 1.1 g/mL
Limit of free ethylene oxide (Target <1 ppm) For information only
Total microbial count (No growth detected) Passes Test
Endotoxin For information only
Protein reactive impurities (100x Diluted) For information only
Arsenic (As) For information only
Cadmium (Cd) For information only
Lead (Pb) For information only
Mercury (Hg) For information only
Cobalt (Co) For information only
Nickel (Ni) For information only
Vanadium (V) For information only
Silver (Ag) For information only
Gold (Au) For information only
Iridium (Ir) For information only
Osmium (Os) For information only
Palladium (Pd) For information only
Platinum (Pt) For information only
Rhodium (Rh) For information only
Ruthenium (Ru) For information only
Selenium (Se) For information only
Thallium (Tl) For information only
Barium (Ba) For information only
Chromium (Cr) For information only
Copper (Cu) For information only
Lithium (Li) For information only
Molybdenum (Mo) For information only
Antimony (Sb) For information only
Tin (Sn) For information only


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR