Gas burette, Bunte, Class A
Lieferant: witeg Labortechnik
2.970.000
WITG2.970.000EA 599 CHF
WITG2.970.000
Gas burette, Bunte, Class A
Büretten
Gas burette made of borosilicate glass, accuracy Class A.
  • With resistant DIFFICO graduation
  • Main point ring graduation
ST-stopcock with double bore and screw-thread retaining nut.
Order Now
TECHNISCHE DATEN
Environmentally Preferable