Condensers, Allihn, for Soxleth extractors
Lieferant: witeg Labortechnik
2.200.034 2.200.045 2.200.060 2.200.029
WITG2.200.034EA 209 CHF
WITG2.200.034 WITG2.200.045 WITG2.200.060 WITG2.200.029
Condensers, Allihn, for Soxleth extractors
Kühler
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. With NS cone. Water connection with glass nozzles.