Condensers, Allihn
Lieferant: witeg Labortechnik
1.720.250 1.720.400 1.723.250 1.724.250 1.727.400 1.724.400 1.728.400
WITG1.720.250EA 155 CHF
WITG1.720.250 WITG1.720.400 WITG1.723.250 WITG1.724.250 WITG1.727.400 WITG1.724.400 WITG1.728.400
Condensers, Allihn
Kühler
Borosilicate glass 3,3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. With or without ground joints. Water connection with glass nozzles.