Splash heads, bent, acc. to Stutzer
Lieferant: witeg Labortechnik
1.425.005
WITG1.425.005EA 184 CHF
WITG1.425.005
Splash heads, bent, acc. to Stutzer
Destillationsbauteile Destillieraufsätze
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Two standard ground cones, inclined at an angle of 75°. Placed directly on the distillation flask to intercept droplets.