Distilling bridges
Lieferant: witeg Labortechnik
1.200.300 1.200.200
WITG1.200.300EA 126 CHF
WITG1.200.300 WITG1.200.200
Distilling bridges
Destillationsbauteile Destillierbrücken
Borosilicate glass 3,3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Distilling links without condenser, with two vertical NS cones.