Schlenk sample tube, SP Wilmad-LabGlass
Lieferant: SP Industries
LG-10800-100
WILMLG-10800-100EA 414 CHF
WILMLG-10800-100
Schlenk sample tube, SP Wilmad-LabGlass
Röhrchen Reagenzgläser- und Probenröhrchen
Tube incorporates a grease-free high vacuum 0 to 4 mm valve and is suited for pressure/vacuum applications. Ideal for reaction and storage of air-sensitive compounds. Sidearm can be connected to inert atmosphere/vacuum source or a manifold.