Water Softener, Envi GreenSOFT

Lieferant: ENVIROFALK PHARMAWATERSYSTEMS
199234
WERN199234EA 1880 CHF
WERN199234
Water Softener, Envi GreenSOFT
Wasseraufbereitungssysteme Ionenaustauscher
Ready-to-connect systems for the production of soft water.

  • Space-saving installation through cabinet design
  • Integrated GRP cartridge, brine tank, brine valve & water meter
  • Including electronic central control valve for quantity or time-controlled regeneration
  • Menu-driven programming with four selectable languages (DE, FR, EN, ES)

Diagnostic mode and history memory with battery-buffered storage of settings and values in case of power failure. Changeover contact for switching off a reverse osmosis system during regeneration.
New Product
Order Now

TECHNISCHE DATEN

Environmentally Preferable
Nutzinhalt 16000 L
Max. Flussrate 0,6 m³/h
Max. Arbeitsdruck 2,5 - 6 bar
Ein-/Auslass Verbindung ³/₄" AG
Stromzufuhr 230 V/50 Hz
B×T×H 230×400×550 mm
Gewicht 10 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR