VWR®, Flaschen zur Wasserprobenahme, steril
Lieferant: VWR Collection
331-0256EA 403 CHF
331-0256 331-0275 331-0277 VWRI331-0256.1 VWRI331-0267.1 VWRI331-0268.1 VWRI331-0269.1 VWRI331-0270.1 VWRI331-0271.1 VWRI331-0272.1 VWRI331-0273.1 VWRI331-0274.1 VWRI331-0275.1 VWRI331-0276.1 VWRI331-0277.1
VWR®, Flaschen zur Wasserprobenahme, steril
Flaschen Flaschen zur Wasserprobenahme
PET, rein, lebensmitteltauglich, mit PP-Verschlüssen.
  • Different coloured caps for easy indentification of test
  • Tamper-evident strip to ensure sampling integrity
  • Full traceability, batch number and individual bottle number on each label
  • Fully recyclable
  • Cap diameter: 32 mm
Lightweight, durable bottles. Caps designed to shield neck from contamination. Choice of dosed and undosed bottles. Rectangular bottles for better storage with limited space.
Blauer Verschluss: Für natürliches, nicht chloriertes Wasser ohne Natriumthiosulfat.
Roter Verschluss:More Product Information