PharMed® BPT Tubings
Lieferant: Saint Gobain Life Sciences
R6512-36BPT 070539-13 070543-18
VERNR6512-36BPTEA 611 CHF
VERNR6512-36BPT VERN070539-13 VERN070543-18
PharMed® BPT Tubings
Schläuche Schläuche für Peristaltikpumpen
PharMed® BPT Tubing has been formulated to withstand the rigors of peristaltic pumping action, while providing the biocompatible fluid surface required in sensitive bioprocess applications. With its superior flex life characteristics, PharMed BPT tubing simplifies biopharmaceutical manufacturing processes by reducing production downtime due to pump tubing failure.
  • Outlasts silicone tubing in peristaltic pumps by up to 30 times
  • Low particulate spallation
  • Can be autoclaved repeatedly
  • Withstands repeated CIP and SIP cleaning and sterilisation