Pen light
Lieferant: VARTA GERAETEBATTERIE
16611101421
VART16611101421EA 32.4 CHF
VART16611101421
Pen light
Beleuchtung Taschenlampen
This pen light is suitable for everyday situations whenever you need a flashlight.
  • Aluminium housing
  • Practical fastening clip
  • Long light duration by white 5 mm LED
Lieferumfang: Supplied with a high energy AAA battery.