J&W PLOT PT GC columns
Lieferant: VARIAN
CP7565PT CP7536PT CP7539PT CP7538PT CP7518PT CP7515PT CP7568PT CP7534PT 19095P-UO4PT
VARICP7565PTEA 3190 CHF
VARICP7565PT VARICP7536PT VARICP7539PT VARICP7538PT VARICP7518PT VARICP7515PT VARICP7568PT VARICP7534PT HEWL19095P-UO4PT
J&W PLOT PT GC columns
Chromatographiesäulen
PLOT PT columns have an integral particle trap at each end, to eliminate the bother of connecting separate traps, and enable GC/MS with leak-free operation and no anxiety.
  • Enhance analytical capability in the lab using PLOT analyses with MS detection and Capillary Flow Technology (CFT)
  • No more hassles with unions or downstream filters: Our integrated particle trapping technology is incorporated into the column as one continuous length of fused silica tubing
  • Higher throughput: Reproducible flow restriction and increased stability reduce the need for method/
More Product Information