Diatomaceous earth sorbents, Chem Elut™ and Hydromatrix™
Lieferant: VARIAN
12198007 12198008 12198006 12198001 12198002 12198004 12198009 12198011 12198012 12198003 65401507 12198017 12198005 12198010 12198015 12198014 75430260 75530260
VARI12198007EA 1090 CHF
VARI12198007 VARI12198008 VARI12198006 VARI12198001 VARI12198002 VARI12198004 VARI12198009 VARI12198011 VARI12198012 VARI12198003 VARI65401507 VARI12198017 VARI12198005 VARI12198010 VARI12198015 VARI12198014 VARI75430260 VARI75530260
Diatomaceous earth sorbents, Chem Elut™ and Hydromatrix™
Chromatographiesäulen
Chem Elut sorbent for rapid, general sample preparation of biological samples such as plasma, serum, whole blood and urine. Chem Elut products are available in buffered and unbuffered formats. The buffered devices can be used for simple scrubbing operations on organic reactions. The base-treated cartridge can remove residual acid compounds from a variety of matrices.
  • High purity sorbent supported liquid extraction (SLE) applications
  • Delivers excellent tube-to-tube reproducibility
Tox Elut cartridges are effective for forensic analysis of drugs inMore Product Information