Venlafaxine-D6 hydrochloride solution, certified reference material, Cerilliant®, Supelco®
Lieferant: Merck
V-009-1ML
V-009-1MLEA 284 CHF
V-009-1ML
Venlafaxine-D6 hydrochloride solution, certified reference material, Cerilliant®, Supelco®
Standards Organische Standards
A new internal standard suitable for many LC/MS and GC/MS applications from forensic or clinical toxicology analysis to urine drug testing and isotope dilution methods. 
Also known by the brand name Effexor®, venlafaxine is an SNRI antidepressant approved for the treatment of major depressive and general anxiety disorders.
Verpackung: Snap-N-Spike®/Snap-N-Shoot®
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5c9b1e5de28d17fb1976aab726ff95dbf32ab6a&vid=d579d74e74e8aa241c9b218155869615bab192fc&prid=web&clid=SAPDAM