Radiometers, digital, UVX
Lieferant: UVP ULTRA VIOLET PRODUCTS
97-0015-02
UVPA97-0015-02EA 1520 CHF
UVPA97-0015-02
Radiometers, digital, UVX
Belichtungsmesser
This digital UVX Radiometer is used with one of the three interchangeable sensors (ordered separately) for measuring 254, 365 and 302 nm UV wavelengths. Measurement of lamp irradiance is the most effective way to monitor the useful life of any UV light source.
  • Order from three sensors: UVX-25 shortwave, UVX-31 midrange or UVX-36 longwave
  • The chart recorder port, standard on the UVX, allows for continuous monitoring
  • The UVX Radiometer is powered by a 9-volt battery for up to 120 hours
  • Unit includes a port for attachment of a chart recorder (user provided)
More Product Information