Anti-USP28 Rabbit Polyclonal Antibody

Lieferant: US Biological
U4101-28D
USBIU4101-28DEA 937 CHF
USBIU4101-28D
Anti-USP28 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: USP28 (ubiquitin specific peptidase 28)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR