Anti-UBE2C Rabbit Polyclonal Antibody

Lieferant: US Biological
U1000-86C
USBIU1000-86CEA 937 CHF
USBIU1000-86C
Anti-UBE2C Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: UBE2C (ubiquitin-conjugating enzyme E2C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

TECHNISCHE DATEN

Antigen-Symbol UBE2C
Environmentally Preferable
Antigen-Name Ubiquitin-conjugating enzyme E2C
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Unconjugated
Reaktivität Human
Wirt Rabbit
Isotyp IgG
Western blot Yes
ELISA Yes
Epitop N-terminal
Form liquid
Reinigung purified by ammonium sulfate precipitation
Lagerungspuffer PBS, 0,09% sodium azide.
Lagertemperatur Cool
Transporttemperatur Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR