Anti-TRIM25 Rabbit Polyclonal Antibody

Lieferant: US Biological
T8466-25A
USBIT8466-25AEA 985 CHF
USBIT8466-25A
Anti-TRIM25 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 10-20 ?g/ml

Type: Primary
Antigen: TRIM25 (tripartite motif containing 25)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR