Anti-TGF beta-induced Factor Chicken Polyclonal Antibody

Lieferant: US Biological
T3630-01
USBIT3630-01EA 1030 CHF
USBIT3630-01
Anti-TGF beta-induced Factor Chicken Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: TGF beta-induced Factor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR