Anti-Tetracycline Sheep Polyclonal Antibody

Lieferant: US Biological
T2965-01C
USBIT2965-01CEA 1070 CHF
USBIT2965-01C
Anti-Tetracycline Sheep Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 20 ?g/ml

Type: Primary
Antigen: Tetracycline
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR