Anti-TAG 72 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 0.N.561A]

Lieferant: US Biological
T0710-07B
USBIT0710-07BEA 530 CHF
USBIT0710-07B
Anti-TAG 72 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 0.N.561A]
Antikörper
Recommended dilution: Western Blot: 1-2?g/ml; Immunoprecipitation: 2?g/mg; Immunochemistry: 0.2-0.4?g/ml

Type: Primary
Antigen: TAG 72
Clonality: monoclonal
Clone: 0.N.561A
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Bovine, Dog, Human, Rat
Order Now

TECHNISCHE DATEN

Antigen-Symbol TAG 72
Environmentally Preferable
Antikörper-Typ Primär
Klonalität Monoklonal
Konjugation HRP (Horseradish Peroxidase)
Klon 0.N.561A
Reaktivität Human,Bovine,Dog,Rat
Wirt Mouse
Isotyp IgG1 kappa
Western blot Yes
Immunpräzipitation Yes
Durchflusszytometrie Yes
Form liquid
Reinigung purified by Protein G chromatography
Lagertemperatur Cool
Transporttemperatur Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR