Anti-SYP Mouse Monoclonal Antibody [clone: 10K361]

Lieferant: US Biological
S9108-07A
USBIS9108-07AEA 757 CHF
USBIS9108-07A
Anti-SYP Mouse Monoclonal Antibody [clone: 10K361]
Antikörper
Recommended dilution: Western Blot: 0.5-1 ?g/ml; Immunochemistry: 1-2?g/ml

Type: Primary
Antigen: SYP (synaptophysin)
Clonality: Monoclonal
Clone: 10K361
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR