Anti-SEF Rabbit Polyclonal Antibody (Rhodamine)

Lieferant: US Biological
S0639-05D
USBIS0639-05DEA 1630 CHF
USBIS0639-05D
Anti-SEF Rabbit Polyclonal Antibody (Rhodamine)
Antikörper
Type: Primary
Antigen: SEF
Clonality: Polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR