Anti-Secretory Component Goat Polyclonal Antibody (Rhodamine)

Lieferant: US Biological
S0630-01P
USBIS0630-01PEA 1050 CHF
USBIS0630-01P
Anti-Secretory Component Goat Polyclonal Antibody (Rhodamine)
Antikörper
Type: Primary
Antigen: Secretory Component
Clonality: Polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Monkey
Order Now

TECHNISCHE DATEN

Antigen-Symbol Secretory Component
Environmentally Preferable
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation TRITC (Tetramethylrhodamine Isothiocyanate)
Reaktivität Monkey
Wirt Goat
Isotyp IgG
ELISA Yes
Form lyophilized powder
Reinigung purified by DEAE chromatography
Lagerungspuffer PBS, pH 7,2 Labled Rhodamine. 1ml sterile ddH2O. Dilute further
Lagertemperatur Cool
Transporttemperatur Ice
Validierte Anwendungen Fluoresence ELISA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR